ContactUsLinkComponent
This product is prepared by labeling high quality goat anti-mouse IgG (H+L) with alkaline phosphatase (AP). AP conjugates could be used for Western Blot, ELISA, immunohistochemistry and other standard immunoassay applications. 1 * 100 µl
Catalog # 20464-100UL
Supplier:  Biotium
This product is prepared by labeling high quality goat anti-mouse IgG (H+L) with alkaline phosphatase (AP). AP conjugates could be used for Western Blot, ELISA, immunohistochemistry and other standard immunoassay applications. 1 * 100 µl
Catalog # 20464-100UL
Supplier:  Biotium