ContactUsLinkComponent
This product is prepared by single labeling (DOL=1) highly cross-adsorbed goat anti-rabbit IgG (H+L) with near-IR fluorescent dye CF ™750ST for (d)-STORM super-resolution microscopy. To minimize cross-reactivity, the antibody has been adsorbed agai e 1 * 500 µl
Catalog # 20826-500UL
Supplier:  Biotium
This product is prepared by single labeling (DOL=1) highly cross-adsorbed goat anti-rabbit IgG (H+L) with near-IR fluorescent dye CF ™750ST for (d)-STORM super-resolution microscopy. To minimize cross-reactivity, the antibody has been adsorbed agai e 1 * 500 µl
Catalog # 20826-500UL
Supplier:  Biotium