ContactUsLinkComponent
Vane probe Ø 100 mm, for measurements at duct outlet Measuring range: +0.10 # + 15 m/s 0# +60 °C Accuracy: ± (0.1 m/s +1.5% V. Mw.) ±0.5 °C Resolution: 0.01 m/s 0.1 °C 1 * 1 items
Catalog # TEST0635.9343
Supplier:  Testo
Vane probe Ø 100 mm, for measurements at duct outlet Measuring range: +0.10 # + 15 m/s 0# +60 °C Accuracy: ± (0.1 m/s +1.5% V. Mw.) ±0.5 °C Resolution: 0.01 m/s 0.1 °C 1 * 1 items
Catalog # TEST0635.9343
Supplier:  Testo
Frequently Bought Together