Order Entry
Northern Ireland
ContactUsLinkComponent
RAT E-CADHERIN ELISA KIT SIMPLESTEP 1 * 96 Tests
Rat E-Cadherin ELISA Kit
Catalog # ABCAAB202413-96
Supplier:  Abcam
CAS Number:  
undefined
Rat E-Cadherin ELISA Kit
Catalog # ABCAAB202413-96
Supplier:  Abcam
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Target protein:
    E Cadherin
  • Description:
    Rat E-Cadherin ELISA kit
  • Sample type:
    Cell culture supernatant, citrate plasma, edta plasma, heparin plasma, serum, urine
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Detection range:
    156,25 - 10000 pg/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    17 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Rat E-Cadherin ELISA kit is a single-wash 90-min Simplestep used to quantify Rat E-Cadherin with a sensitivity of 17 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.

Frequently Bought Together