ContactUsLinkComponent
RS-485 interface (compatible with 1535 Monitor/Alarm only) Data Output, For use with 3962 Environmental Chamber/Incubator 1 * 1 items
Catalog # HERA190523
Supplier:  Thermo Scientific
RS-485 interface (compatible with 1535 Monitor/Alarm only) Data Output, For use with 3962 Environmental Chamber/Incubator 1 * 1 items
Catalog # HERA190523
Supplier:  Thermo Scientific
Frequently Bought Together