ContactUsLinkComponent
SIEVE ISO3310-1 200x50MM 5.60MM SS PLATE 1 * 1 items
Catalog # ENDE200SIS5.60
Supplier:  ENDECOTTS
SIEVE ISO3310-1 200x50MM 5.60MM SS PLATE 1 * 1 items
Catalog # ENDE200SIS5.60
Supplier:  ENDECOTTS
Frequently Bought Together