FLUORESCEIN - CONJUGATED GOAT IgG FRACTION TO RABBIT IgG (WHOLE MOLECULE). Anti-Rabbit Host: goat Form: lyophilized (IgG) Applicatio ns: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Conjugated 1 * 2 mL
Catalog # ICNA0855646
Supplier: MP Biomedicals