ContactUsLinkComponent
RABBIT IgG FRACTION TO CHICKEN RED BLOOD CELLS. Anti-ChickenHost: rabbitForm: lyophilized (IgG)Applications: Immunoblotting Immunos taining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 5 mL
Catalog # ICNA0855306
Supplier:  MP Biomedicals
RABBIT IgG FRACTION TO CHICKEN RED BLOOD CELLS. Anti-ChickenHost: rabbitForm: lyophilized (IgG)Applications: Immunoblotting Immunos taining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 5 mL
Catalog # ICNA0855306
Supplier:  MP Biomedicals
Frequently Bought Together