FLUORESCEIN - CONJUGATED GOAT IgG FRACTION TO CAT IgG (WHOLE MOLECULE). Anti-Cat Host: goat Form: lyophilized (IgG) Applications: Im munostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Conjugated 1 * 2 mL
Catalog # ICNA0855291
Supplier: MP Biomedicals