ContactUsLinkComponent
FITC CONJUGATED GOAT AFFINITY PURIFIED IgG TO RAT IgG (WHOLE MOLECULE) (NO CROSS HUMAN). Anti-RatHost: goatForm: affinity purified l iquidApplications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections.This 1 * 2 mL
Catalog # ICNA0856407
Supplier:  MP Biomedicals
FITC CONJUGATED GOAT AFFINITY PURIFIED IgG TO RAT IgG (WHOLE MOLECULE) (NO CROSS HUMAN). Anti-RatHost: goatForm: affinity purified l iquidApplications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections.This 1 * 2 mL
Catalog # ICNA0856407
Supplier:  MP Biomedicals