ContactUsLinkComponent
GELLAN GUM POWDER. Agar substitute gelling agent. Natural gelling agent used to produce clear media for microbiological and other ap plication. 1 * 1 kg
Catalog # ICNA0218010601
Supplier:  MP Biomedicals
GELLAN GUM POWDER. Agar substitute gelling agent. Natural gelling agent used to produce clear media for microbiological and other ap plication. 1 * 1 kg
Catalog # ICNA0218010601
Supplier:  MP Biomedicals
Frequently Bought Together