ContactUsLinkComponent
Fast Flow Free-flowing powder for column chromatography. This unique form of calcium phosphate has an ino rganic matrix ideal for fractionation and purification of proteins, enzymes, steroids, nucleic acids, plasmids and viruses. 1 * 10 g
Catalog # ICNA0215016210
Supplier:  MP Biomedicals
Fast Flow Free-flowing powder for column chromatography. This unique form of calcium phosphate has an ino rganic matrix ideal for fractionation and purification of proteins, enzymes, steroids, nucleic acids, plasmids and viruses. 1 * 10 g
Catalog # ICNA0215016210
Supplier:  MP Biomedicals