ContactUsLinkComponent
From Porcine Heart Lyophilized powder. Activity: Approx. 80 units/mg protein. Unit Definition: One unit will cause the transamination of 1.0 µ mole of L-alanine per minute at pH 7 and 25°C. Contaminants: GLDH, GOT, LDH, MDH: 0.01% . 1 * 1 KU
Catalog # ICNA0215119191
Supplier:  MP Biomedicals
From Porcine Heart Lyophilized powder. Activity: Approx. 80 units/mg protein. Unit Definition: One unit will cause the transamination of 1.0 µ mole of L-alanine per minute at pH 7 and 25°C. Contaminants: GLDH, GOT, LDH, MDH: 0.01% . 1 * 1 KU
Catalog # ICNA0215119191
Supplier:  MP Biomedicals