ContactUsLinkComponent
Crystalline Useful pH range 7.3-8.7 This is the propane analog of HEPES. 1 * 250 g
Catalog # ICNA0210192783
Supplier:  MP Biomedicals
Crystalline Useful pH range 7.3-8.7 This is the propane analog of HEPES. 1 * 250 g
Catalog # ICNA0210192783
Supplier:  MP Biomedicals
Frequently Bought Together