ContactUsLinkComponent
1-NAPHTHYL PHOSPHATE MONOSODIUM SALT HYDRATE. A non-specific phosphatase inhibitor. Inhibits acid, alkaline and protein phosphatases . 1 * 5 g
Catalog # ICNA0219941005
Supplier:  MP Biomedicals
1-NAPHTHYL PHOSPHATE MONOSODIUM SALT HYDRATE. A non-specific phosphatase inhibitor. Inhibits acid, alkaline and protein phosphatases . 1 * 5 g
Catalog # ICNA0219941005
Supplier:  MP Biomedicals
Frequently Bought Together