ContactUsLinkComponent
Rat or Mouse Powdered,AIN-76A diet with 40% corn starch and sucrose. 1 * 20 kg
Catalog # ICNA0296035420
Supplier:  MP Biomedicals
Rat or Mouse Powdered,AIN-76A diet with 40% corn starch and sucrose. 1 * 20 kg
Catalog # ICNA0296035420
Supplier:  MP Biomedicals