SODIUM PERCHLORATE MONOHYDRATE ACS REAGENT GRADE. Purity: 85-90% 1 * 500 g
Catalog # ICNA0219486290
Supplier: MP Biomedicals
SODIUM PERCHLORATE MONOHYDRATE ACS REAGENT GRADE. Purity: 85-90% 1 * 500 g
Catalog # ICNA0219486290
Supplier: MP Biomedicals
Frequently Bought Together
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Dimethyl sulfoxide EMPLURA 1 * 185 L
Supplier: Merck
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Hexane (mixture of isomers) 99% ACS
Supplier: Thermo Fisher Scientific
Cyclohexane for pesticide residue analysis
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific