ContactUsLinkComponent
MES MONOHYDRATE FREE ACID. Molecular Biology Reagent. Purity: >99% Zwitterionic bufferpKa = 6.15 at 25C 1 * 10 g
Catalog # ICNA0219483510
Supplier:  MP Biomedicals
MES MONOHYDRATE FREE ACID. Molecular Biology Reagent. Purity: >99% Zwitterionic bufferpKa = 6.15 at 25C 1 * 10 g
Catalog # ICNA0219483510
Supplier:  MP Biomedicals
Frequently Bought Together