ContactUsLinkComponent
FORSKOLIN. From Coleus forskohlii (7#-acetoxy-8,13-epoxy-1a,6# ,9a-trihydroxy-labd-14-ene-11-one)Reported to demonstrate positive ino tropic and heart rate-increasing activity at low doses in animals. 1 * 25 mg
Catalog # ICNA0219066925
Supplier:  MP Biomedicals
FORSKOLIN. From Coleus forskohlii (7#-acetoxy-8,13-epoxy-1a,6# ,9a-trihydroxy-labd-14-ene-11-one)Reported to demonstrate positive ino tropic and heart rate-increasing activity at low doses in animals. 1 * 25 mg
Catalog # ICNA0219066925
Supplier:  MP Biomedicals