ContactUsLinkComponent
PROTEIN S. From Human PlasmaPurified protein containing glycine and NaCl.A vitamin K dependent protein which is a cofactor of activa ted protein C (APC). It increases the affinity of APC for phospholipids which leads to factor V and VIII inhibition. 1 * 100 µG
Catalog # ICNA0219408180
Supplier:  MP Biomedicals
PROTEIN S. From Human PlasmaPurified protein containing glycine and NaCl.A vitamin K dependent protein which is a cofactor of activa ted protein C (APC). It increases the affinity of APC for phospholipids which leads to factor V and VIII inhibition. 1 * 100 µG
Catalog # ICNA0219408180
Supplier:  MP Biomedicals