ContactUsLinkComponent
YEAST EXTRACT. Molecular Biology ReagentA vacuum-dried extract concentrate of Baker's yeast containing the B-complex factors of appr oximately three times the weight of ordinary dry yeast. An alternative to beef extract for general bacteriological us 1 * 1 kg
Catalog # ICNA0219402791
Supplier:  MP Biomedicals
YEAST EXTRACT. Molecular Biology ReagentA vacuum-dried extract concentrate of Baker's yeast containing the B-complex factors of appr oximately three times the weight of ordinary dry yeast. An alternative to beef extract for general bacteriological us 1 * 1 kg
Catalog # ICNA0219402791
Supplier:  MP Biomedicals
Frequently Bought Together