ContactUsLinkComponent
USP Grade.Purity: 99.95% min.Buffering pH range 7.0-9.0 pKa at 25C =8.1. Excellent biochemical and biological buffer for all applications where ultra high purity is required. 1 * 100 g
Catalog # ICNA0219560580
Supplier:  MP Biomedicals
USP Grade.Purity: 99.95% min.Buffering pH range 7.0-9.0 pKa at 25C =8.1. Excellent biochemical and biological buffer for all applications where ultra high purity is required. 1 * 100 g
Catalog # ICNA0219560580
Supplier:  MP Biomedicals
Frequently Bought Together