ContactUsLinkComponent
METHYLENE BLUE TRIHYDRATE. Zinc FreeTrihydrateMolecular Biology Reagent.An ethidium bromide alternative for the visualization of nucleic acids in gels.Gels are viewable in visible light.Does not require a darkroom.No UV induced DNA damage Gels can be 1 * 5 g
Catalog # ICNA0219399805
Supplier:  MP Biomedicals
METHYLENE BLUE TRIHYDRATE. Zinc FreeTrihydrateMolecular Biology Reagent.An ethidium bromide alternative for the visualization of nucleic acids in gels.Gels are viewable in visible light.Does not require a darkroom.No UV induced DNA damage Gels can be 1 * 5 g
Catalog # ICNA0219399805
Supplier:  MP Biomedicals
Frequently Bought Together