ContactUsLinkComponent
L-ORNITHINE HYDROCHLORIDE. Cell Culture Reagent Hydrochloride Crystalline. Purity: 99% . This material is essentially free of citrulline and ammonia. 1 * 100 g
Catalog # ICNA0219471880
Supplier:  MP Biomedicals
L-ORNITHINE HYDROCHLORIDE. Cell Culture Reagent Hydrochloride Crystalline. Purity: 99% . This material is essentially free of citrulline and ammonia. 1 * 100 g
Catalog # ICNA0219471880
Supplier:  MP Biomedicals
Frequently Bought Together