ContactUsLinkComponent
PANCREATIN 8X USP GRADE. From Porcine Pancreas USP Grade Reagent Composed of numerous enzymes including amylase, trypsin, lipase, ribo nuclease and protease. 1 * 100 g
Catalog # ICNA0219397680
Supplier:  MP Biomedicals
PANCREATIN 8X USP GRADE. From Porcine Pancreas USP Grade Reagent Composed of numerous enzymes including amylase, trypsin, lipase, ribo nuclease and protease. 1 * 100 g
Catalog # ICNA0219397680
Supplier:  MP Biomedicals
Frequently Bought Together