ContactUsLinkComponent
5 - BROMO - 4 - CHLORO - 3 - INDOLYL - alpha - D - GALACTOPYRANOSIDE. Purity: >98% An #-galactoside substrate which differentiates # -galactosidase positive yeast strains or bacteria. Protect from light and humidity. 1 * 100 mg
Catalog # ICNA0219392180
Supplier:  MP Biomedicals
5 - BROMO - 4 - CHLORO - 3 - INDOLYL - alpha - D - GALACTOPYRANOSIDE. Purity: >98% An #-galactoside substrate which differentiates # -galactosidase positive yeast strains or bacteria. Protect from light and humidity. 1 * 100 mg
Catalog # ICNA0219392180
Supplier:  MP Biomedicals