D-(+)-GALACTOSE ANHYDROUS. Cell Culture Reagent.Anhydrous Crystalline Purity: 98% 1 * 100 g
Catalog # ICNA0219467080
Supplier: MP Biomedicals
D-(+)-GALACTOSE ANHYDROUS. Cell Culture Reagent.Anhydrous Crystalline Purity: 98% 1 * 100 g
Catalog # ICNA0219467080
Supplier: MP Biomedicals
Frequently Bought Together
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
2-Propanol, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
N,N-Dimethylformamide ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane ≥99%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Cyclohexane, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
Solvent ASTM D2896, Chlorobenzene/Acetic acid-mixture (2/1 v/v), TBN solvent, VOLUSOL®
Supplier: VWR Chemicals
Methanol, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Chloroform ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethylene glycol monomethyl ether (2-methoxyethanol), extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Phytanic acid solution ≥96% in ethanol
Supplier: Cayman Chemical
Glycerine, anhydrous ≥99.5% (by GC), Puriss. p.a. ACS, distilled, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane ≥99.9% stabilised, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals