CALCIUM NITRATE TETRAHYDRATE ACS REAGENT GRADE.Purity: >99% 1 * 250 g
ICNA0219380183
CALCIUM NITRATE TETRAHYDRATE ACS REAGENT GRADE.Purity: >99% 1 * 250 g
ICNA0219380183
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals