ContactUsLinkComponent
Purity: >98% Present in wine and grapes and found to reduce serum lipids, inhibit platelet aggregation and act as a chemopreventative agent and antioxidant. 1 * 5 mg
Catalog # ICNA0219605205
Supplier:  MP Biomedicals
Purity: >98% Present in wine and grapes and found to reduce serum lipids, inhibit platelet aggregation and act as a chemopreventative agent and antioxidant. 1 * 5 mg
Catalog # ICNA0219605205
Supplier:  MP Biomedicals
Frequently Bought Together