From Canavalia ensiformis Highly Purifed Esssentially Salt Free Possesses specificity for terminal -D -mannosyl and -D -glucosyl residues and also agglutinates red blood cells. Caution: Has mitogenic properties. 1 * 1 g
Catalog # ICNA0219528301
Supplier: MP Biomedicals