ContactUsLinkComponent
From Canavalia ensiformis Highly Purifed Esssentially Salt Free Possesses specificity for terminal -D -mannosyl and -D -glucosyl residues and also agglutinates red blood cells. Caution: Has mitogenic properties. 1 * 50 mg
Catalog # ICNA02195283.6
Supplier:  MP Biomedicals
From Canavalia ensiformis Highly Purifed Esssentially Salt Free Possesses specificity for terminal -D -mannosyl and -D -glucosyl residues and also agglutinates red blood cells. Caution: Has mitogenic properties. 1 * 50 mg
Catalog # ICNA02195283.6
Supplier:  MP Biomedicals
Frequently Bought Together