ContactUsLinkComponent
PUROMYCIN DIHYDROCHLORIDE. Cell Culture Reagent Dihydrochloride Crystalline Antimicrobial Causes premature chain termination in protein synthesis, and is an inhibitor of aminopeptidase and enkephalinase. 1 * 100 mg
Catalog # ICNA0219453980
Supplier:  MP Biomedicals
PUROMYCIN DIHYDROCHLORIDE. Cell Culture Reagent Dihydrochloride Crystalline Antimicrobial Causes premature chain termination in protein synthesis, and is an inhibitor of aminopeptidase and enkephalinase. 1 * 100 mg
Catalog # ICNA0219453980
Supplier:  MP Biomedicals
Frequently Bought Together