PUROMYCIN DIHYDROCHLORIDE. Cell Culture Reagent Dihydrochloride Crystalline Antimicrobial Causes premature chain termination in protein synthesis, and is an inhibitor of aminopeptidase and enkephalinase. 1 * 10 mg
Catalog # ICNA0219453910
Supplier: MP Biomedicals
Frequently Bought Together