ContactUsLinkComponent
SC 100-S13 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, SC100-S13, Amb+13 to 100 C, 7-11 L BATH VOLUME, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR 1 * 1 items
Catalog # HAKA1521131
SC 100-S13 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, SC100-S13, Amb+13 to 100 C, 7-11 L BATH VOLUME, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR 1 * 1 items
Catalog # HAKA1521131
Frequently Bought Together