ContactUsLinkComponent
A conventional antipsychotic drug that potently activates several neuroreceptors, including serotonin (Kis = 10, 60, and 57 nM for 5 -HT2A, 5-HT2C, and 5-HT6, respectively), dopamine D2 (Ki = 11 nM), and adrenergic 1A (Ki = 5 nM) receptors 1 * 25 g
Catalog # CAYM14400-25
Supplier:  Cayman Chemical
A conventional antipsychotic drug that potently activates several neuroreceptors, including serotonin (Kis = 10, 60, and 57 nM for 5 -HT2A, 5-HT2C, and 5-HT6, respectively), dopamine D2 (Ki = 11 nM), and adrenergic 1A (Ki = 5 nM) receptors 1 * 25 g
Catalog # CAYM14400-25
Supplier:  Cayman Chemical
Frequently Bought Together