ContactUsLinkComponent
A chromogenic substrate for N-acetyl-D-galactosaminidase that generates a yellow solution upon cleavage 1 * 25 mg
Catalog # CAYM16755-25
Supplier:  Cayman Chemical
A chromogenic substrate for N-acetyl-D-galactosaminidase that generates a yellow solution upon cleavage 1 * 25 mg
Catalog # CAYM16755-25
Supplier:  Cayman Chemical