A reversible inhibitor of the auto and transphosphorylation kinase activity of the erbB receptor tyrosine kinase superfamily: EGFR ( IC50 = 8 µM), erbB-2 (IC50 = 49 nM), erbB3, and erbB4 (IC50 = 52 nM) 1 * 25 mg
Catalog # CAYM13329-25
Supplier: Cayman Chemical