ContactUsLinkComponent
A potent inhibitor of IB kinases (IKKs), displaying IC50 values of 200, 40, and 70 nM for IKK, IKK, and IKK complex, respectively , in cell-free assays 1 * 5 mg
Catalog # CAYM13313-5
Supplier:  Cayman Chemical
A potent inhibitor of IB kinases (IKKs), displaying IC50 values of 200, 40, and 70 nM for IKK, IKK, and IKK complex, respectively , in cell-free assays 1 * 5 mg
Catalog # CAYM13313-5
Supplier:  Cayman Chemical