ContactUsLinkComponent
A conjugate of CNP and -D-glucose, to be used as a chromogenic substrate for enzymes that target the linkage between CNP and -D-gl ucose 1 * 50 mg
Catalog # CAYM16527-50
Supplier:  Cayman Chemical
A conjugate of CNP and -D-glucose, to be used as a chromogenic substrate for enzymes that target the linkage between CNP and -D-gl ucose 1 * 50 mg
Catalog # CAYM16527-50
Supplier:  Cayman Chemical