ContactUsLinkComponent
A tricyclic antidepressant that potently inhibits serotonin and norepinephrine reuptake (Kis = 7.4 and 96 nM, respectively) 1 * 1 g
Catalog # CAYM15884-1
Supplier:  Cayman Chemical
A tricyclic antidepressant that potently inhibits serotonin and norepinephrine reuptake (Kis = 7.4 and 96 nM, respectively) 1 * 1 g
Catalog # CAYM15884-1
Supplier:  Cayman Chemical
Frequently Bought Together