ContactUsLinkComponent
An ALK1 and ALK2 inhibitor (IC50s = 1.8 and 1.1 nM, respectively) that can prevent BMP4-induced SMAD1/5/8 pathway activation in vitr o (IC50 = 100 nM) without affecting TGF- signaling 1 * 1 mg
Catalog # CAYM16678-1
Supplier:  Cayman Chemical
An ALK1 and ALK2 inhibitor (IC50s = 1.8 and 1.1 nM, respectively) that can prevent BMP4-induced SMAD1/5/8 pathway activation in vitr o (IC50 = 100 nM) without affecting TGF- signaling 1 * 1 mg
Catalog # CAYM16678-1
Supplier:  Cayman Chemical