ContactUsLinkComponent
A potent inhibitor of Aurora kinases (IC50s = 15, 25 and 19 nM for Aurora A, B, and C, respectively) 1 * 10 mg
Catalog # CAYM15552-10
Supplier:  Cayman Chemical
A potent inhibitor of Aurora kinases (IC50s = 15, 25 and 19 nM for Aurora A, B, and C, respectively) 1 * 10 mg
Catalog # CAYM15552-10
Supplier:  Cayman Chemical