ContactUsLinkComponent
An irreversible inhibitor of the fatty acid translocase CD36, blocking uptake of oleate, linoleate, or stearate by about 65% when ad ded at 200 µM to adipocytes 1 * 50 mg
Catalog # CAYM11211-50
Supplier:  Cayman Chemical
An irreversible inhibitor of the fatty acid translocase CD36, blocking uptake of oleate, linoleate, or stearate by about 65% when ad ded at 200 µM to adipocytes 1 * 50 mg
Catalog # CAYM11211-50
Supplier:  Cayman Chemical
Frequently Bought Together