ContactUsLinkComponent
A natural neurotransmitter and hormone that acts as an agonist of adrenergic receptors (Ki values are 330, 56, and 740 nM for 1, 2 , and 1 adrenoceptors, respectively) 1 * 100 mg
Catalog # CAYM16673-100
Supplier:  Cayman Chemical
A natural neurotransmitter and hormone that acts as an agonist of adrenergic receptors (Ki values are 330, 56, and 740 nM for 1, 2 , and 1 adrenoceptors, respectively) 1 * 100 mg
Catalog # CAYM16673-100
Supplier:  Cayman Chemical