ContactUsLinkComponent
A benzazepine derivative that antagonizes the binding of AVP to vasopressin V2 receptors (IC50 = 14 nM) 1 * 100 mg
  CAYM16664-100
A benzazepine derivative that antagonizes the binding of AVP to vasopressin V2 receptors (IC50 = 14 nM) 1 * 100 mg
  CAYM16664-100