ContactUsLinkComponent
A benzazepine derivative that antagonizes the binding of AVP to vasopressin V2 receptors (IC50 = 14 nM) 1 * 10 mg
Catalog # CAYM16664-10
Supplier:  Cayman Chemical
A benzazepine derivative that antagonizes the binding of AVP to vasopressin V2 receptors (IC50 = 14 nM) 1 * 10 mg
Catalog # CAYM16664-10
Supplier:  Cayman Chemical