ContactUsLinkComponent
A pan inhibitor PKD (IC50s = 1, 2.5, and 2 nM for PKD1, PKD2, and PKD3, respectively) 1 * 1 mg
Catalog # CAYM15337-1
Supplier:  Cayman Chemical
A pan inhibitor PKD (IC50s = 1, 2.5, and 2 nM for PKD1, PKD2, and PKD3, respectively) 1 * 1 mg
Catalog # CAYM15337-1
Supplier:  Cayman Chemical