ContactUsLinkComponent
A selective JAK2 inhibitor that blocks signaling (IC50 = 58 nM at 5 mM ATP) and proliferation of JAK2-expressing cell lines (GI50 = 60 nM) 1 * 10 mg
Catalog # CAYM10702-10
Supplier:  Cayman Chemical
A selective JAK2 inhibitor that blocks signaling (IC50 = 58 nM at 5 mM ATP) and proliferation of JAK2-expressing cell lines (GI50 = 60 nM) 1 * 10 mg
Catalog # CAYM10702-10
Supplier:  Cayman Chemical