ContactUsLinkComponent
A selective, reversible inhibitor of MAO-B (Ki = 0.091 M) over MAO-A (Ki = 9.06 M) 1 * 500 mg
Catalog # CAYM15046-500
Supplier:  Cayman Chemical
A selective, reversible inhibitor of MAO-B (Ki = 0.091 M) over MAO-A (Ki = 9.06 M) 1 * 500 mg
Catalog # CAYM15046-500
Supplier:  Cayman Chemical