ContactUsLinkComponent
A cell-permeable inhibitor of Hhat (IC50 = 0.85 µM) 1 * 5 mg
Catalog # CAYM16972-5
Supplier:  Cayman Chemical
A cell-permeable inhibitor of Hhat (IC50 = 0.85 µM) 1 * 5 mg
Catalog # CAYM16972-5
Supplier:  Cayman Chemical